Merck
  • NLRP3 Inflammasome Deficiency Alleviates Inflammation and Oxidative Stress by Promoting PINK1/Parkin-Mediated Mitophagy in Allergic Rhinitis Mice and Nasal Epithelial Cells.
NLRP3 Inflammasome Deficiency Alleviates Inflammation and Oxidative Stress by Promoting PINK1/Parkin-Mediated Mitophagy in Allergic Rhinitis Mice and Nasal Epithelial Cells.
Journal of asthma and allergy (2024-08-06)
Hong Ding, Xiaofan Lu, Huimin Wang, Wenming Chen, Bing Niu
ABSTRACT
Accumulating evidence indicates that oxidative stress and inflammation are the pathological basis of allergic diseases. Inhibition of NOD-like receptor family pyrin domain-containing 3 (NLRP3) inflammasome could ameliorate allergic rhinitis (AR). Here, we explored the effects and mechanisms that underlie NLRP3 inhibition on oxidative stress and inflammation in AR. Ovalbumin (OVA)-induced AR murine model was established using wild-type (WT) and NLRP3-deficient mice. HNEpCs were stimulated with interleukin (IL)-13 with MCC950 pretreatment or PTEN-induced putative kinase 1 (PINK1) siRNA. The indicators of oxidative stress, inflammation, apoptosis, and mitophagy were determined both in vivo and in vitro. NLRP3 knockout (KO) reduced the frequency of nasal rubbing and sneezing, the infiltration of eosinophils, the number of mast cells, and the accumulation of goblet cells in AR mice after OVA stimulation. The NLRP3 KO AR mice exhibited the increased concentrations of OVA-specific immunoglobulin E (OVA-sIgE), IL-1β, IL-4, IL-13, IL-6, TNF-α, and the upregulated level of IFN-γ. NLRP3 KO significantly inhibited oxidative stress, and also markedly decreased apoptosis in the nasal mucosa of AR mice. Moreover, evaluated protein expressions of PINK1, enzyme 3 (E3) ubiquitin ligase PRKN (Parkin), and LC3 II, decreased expression of TOM20, as well as the increased colocalization of LC3 with mitochondria were observed in NLRP3 KO AR mice. In vitro, IL-13 exposure increased the levels of NLRP3 and IL-1β. Inhibition of NLRP3 using MCC950 enhanced PINK1/Parkin-mediated mitophagy but attenuated inflammation, oxidative stress, and apoptosis. However, PINK1 knockdown abrogated mitophagy and also reversed the protective effects of MCC950 on inflammation, oxidative stress, and apoptosis in HNEpCs stimulated with IL-13. Inhibition of NLRP3 inflammasome exerts the protective effects on AR by facilitating mitophagy regulated by PINK1/Parkin signaling pathway.