Merck
  • LSR antibody promotes apoptosis and disrupts epithelial barriers via signal pathways in endometrial cancer.
LSR antibody promotes apoptosis and disrupts epithelial barriers via signal pathways in endometrial cancer.
Tissue barriers (2022-07-28)
Kimihito Saito, Takumi Konno, Takayuki Kohno, Hiroshi Shimada, Motoki Matsuura, Tadahi Okada, Arisa Kura, Daichi Ishii, Masuo Kondoh, Tsuyoshi Saito, Takashi Kojima
ABSTRACT
Lipolysis-stimulated lipoprotein receptor (LSR), a lipid metabolism-related factor localized in tricellular tight junctions (tTJs), plays an important role in maintaining the epithelial barrier. LSR is highly expressed in well-differentiated endometrial endometrioid carcinoma (EEC), and its expression decreases during malignancy. Angubindin-1, a novel LSR ligand peptide, regulates tTJs without cytotoxicity, enhances paracellular permeability, and regulates epithelial barrier via c-Jun N-terminal kinase (JNK)/cofilin. In this study, we investigated the immune-modulatory roles of an anti-LSR antibody in the treatment of EEC in vitro compared to those of angubindin-1. We prepared an antibody against the extracellular N-terminal domain of human LSR (LSR-N-ab) and angubindin-1. EEC cell-line Sawano cells in 2D and 2.5D cultures were treated with 100 μg/ml LSR-N-ab or 2.5 μg/ml angubindin-1 with or without protein tyrosine kinase 2β inhibitor PF431396 (PF43) and JNK inhibitor SP600125 (SP60) at 10 μM. Treatment with LSR-N-ab and angubindin-1 decreased LSR at the membranes of tTJs and the activity of phosphorylated LSR and phosphorylated cofilin in 2D culture. Treatment with LSR-N-ab and angubindin-1 decreased the epithelial barrier measured as TEER values in 2D culture and enhanced the epithelial permeability of FD-4 in 2.5D culture. Treatment with LSR-N-ab, but not angubindin-1, induced apoptosis in 2D culture. Pretreatment with PF43 and SP60 prevented all the changes induced by treatment with LSR-N-ab and angubindin-1. Treatment with LSR-N-ab and angubindin-1 enhanced the cell metabolism measured as the mitochondrial respiration levels in 2D culture. LSR-N-ab and angubindin-1 may be useful for therapy of human EEC via enhanced apoptosis or drug absorption.