Merck
  • TNF and IFNγ-induced cell death requires IRF1 and ELAVL1 to promote CASP8 expression.
TNF and IFNγ-induced cell death requires IRF1 and ELAVL1 to promote CASP8 expression.
The Journal of cell biology (2024-02-06)
Buhao Deng, Jingyi Wang, Tingyun Yang, Zhao Deng, Jiafan Yuan, Bohan Zhang, Zhen Zhou, Fang Chen, Lu Fang, Chengzhi Liang, Bo Yan, Youwei Ai
ABSTRACT
TNFα and IFNγ (TNF/IFNγ) synergistically induce caspase-8 activation and cancer cell death. However, the mechanism of IFNγ in promoting TNF-initiated caspase-8 activation in cancer cells is poorly understood. Here, we found that in addition to CASP8, CYLD is transcriptionally upregulated by IFNγ-induced transcription factor IRF1. IRF1-mediated CASP8 and CYLD upregulation additively mediates TNF/IFNγ-induced cancer cell death. Clinically, the expression levels of TNF, IFNγ, CYLD, and CASP8 in melanoma tumors are increased in patients responsive to immune checkpoint blockade (ICB) therapy after anti-PD-1 treatment. Accordingly, our genetic screen revealed that ELAVL1 (HuR) is required for TNF/IFNγ-induced caspase-8 activation. Mechanistically, ELAVL1 binds CASP8 mRNA and extends its stability to sustain caspase-8 expression both in IFNγ-stimulated and in basal conditions. Consequently, ELAVL1 determines death receptors-initiated caspase-8-dependent cell death triggered from stimuli including TNF and TRAIL by regulating basal/stimulated caspase-8 levels. As caspase-8 is a master regulator in cell death and inflammation, these results provide valuable clues for tumor immunotherapy and inflammatory diseases.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
NU7026, ≥98% (HPLC), solid
Sigma-Aldrich
Cycloheximide, Cycloheximide, CAS 66-81-9, is an antifungal antibiotic that inhibits protein synthesis in eukaryotes but not prokaryotes.
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.