Merck
Smurf1 Modulates Smad Signaling Pathway in Fibrotic Cataract Formation.
Investigative ophthalmology & visual science (2024-02-07)
Fanying Jiang, Yuanfan Yang, Yan Ni, Yingyan Qin, Fa Yuan, Rong Ju, Mingxing Wu
ABSTRACT
TGF-Ī²/BMP signaling pathway plays a significant role in fibrotic cataract. Smurf1, a ubiquitin protein ligase, regulates the TGF-Ī²/BMP signaling pathway through the ubiquitin-proteasome system (UPS). This study aims to investigate the role of Smurf1 in the progression of fibrotic cataract and its underlying mechanism. We used a mouse model of injury-induced anterior subcapsular cataract (ASC) and administered the Smurf1 inhibitor A01 for in vivo investigations. RNA sequencing was performed to examine global gene expression changes. Protein levels were assessed by Simple Western analysis. The volume of subcapsular opacity was determined using whole-mount immunofluorescence of lens anterior capsules. Lentivirus was utilized to establish cell lines with Smurf1 knockdown or overexpression in SRA01/04. Lens epithelial cell (LEC) proliferation was evaluated by CCK8 and EdU assays. Cell cycle profile was determined by flow cytometry. LEC migration was measured using Transwell and wound healing assays. The mRNA levels of genes associated with cell proliferation, migration, epithelial-mesenchymal transition (EMT), TGF-Ī²/BMP pathway, and UPS were upregulated in mouse ASC model. Smurf1 mRNA and protein levels were upregulated in lens capsules of patients and mice with ASC. Anterior chamber injection of A01 inhibited ASC formation and EMT. In vitro, Smurf1 knockdown reduced proliferation, migration and TGF-Ī²2-induced EMT of LECs, concomitant with the upregulation of Smad1, Smad5, and pSmad1/5. Conversely, overexpression of Smurf1 showed opposite phenotypes. Smurf1 regulates fibrotic cataract progression by influencing LEC proliferation, migration, and EMT through the modulation of the Smad signaling pathway, offering a novel target for the fibrotic cataract treatment.
Sigma-Aldrich
A01 (Smurf1 inhibitor), ≥98% (HPLC)