Merck
  • Proinflammatory CD14highCD16low monocytes/macrophages prevail in Treponema phagedenis-associated bovine digital dermatitis.
Proinflammatory CD14highCD16low monocytes/macrophages prevail in Treponema phagedenis-associated bovine digital dermatitis.
Infection and immunity (2024-01-08)
Priyoshi Lahiri, Rakel Arrazuria, Yi Lin Tan, Jeroen De Buck, Morley D Hollenberg, Karin Orsel, Eduardo R Cobo
ABSTRACT
Digital dermatitis (DD) is a skin disease in cattle characterized by painful inflammatory ulcerative lesions in the feet, mostly associated with local colonization by Treponema spp., including Treponema phagedenis. The reason why most DD lesions remain actively inflamed and progress to chronic conditions despite antibiotic treatment remains unknown. Herein, we show an abundant infiltration of proinflammatory (CD14highCD16low) monocytes/macrophages in active DD lesions, a skin response that was not mitigated by topical treatment with oxytetracycline. The associated bacterium, T. phagedenis, isolated from DD lesions in cattle, when injected subcutaneously into mice, induced abscesses with a local recruitment of Ly6G+ neutrophils and proinflammatory (Ly6ChighCCR2+) monocytes/macrophages, which appeared at infection onset (4 days post challenge) and persisted for at least 7 days post challenge. When exploring the ability of macrophages to regulate inflammation, we showed that bovine blood-derived macrophages challenged with live T. phagedenis or its structural components secreted IL-1β via a mechanism dependent on the NLRP3 inflammasome. This study shows that proinflammatory characteristics of monocytes/macrophages and neutrophils dominate active non-healing ulcerative lesions in active DD, thus likely impeding wound healing after antibiotic treatment.