Merck
  • Small extracellular vesicles from hypoxia-preconditioned bone marrow mesenchymal stem cells attenuate spinal cord injury via miR-146a-5p-mediated regulation of macrophage polarization.
Small extracellular vesicles from hypoxia-preconditioned bone marrow mesenchymal stem cells attenuate spinal cord injury via miR-146a-5p-mediated regulation of macrophage polarization.
Neural regeneration research (2024-03-15)
Zeyan Liang, Zhelun Yang, Haishu Xie, Jian Rao, Xiongjie Xu, Yike Lin, Chunhua Wang, Chunmei Chen
ABSTRACT
JOURNAL/nrgr/04.03/01300535-202410000-00027/figure1/v/2024-02-06T055622Z/r/image-tiff Spinal cord injury is a disabling condition with limited treatment options. Multiple studies have provided evidence suggesting that small extracellular vesicles (SEVs) secreted by bone marrow mesenchymal stem cells (MSCs) help mediate the beneficial effects conferred by MSC transplantation following spinal cord injury. Strikingly, hypoxia-preconditioned bone marrow mesenchymal stem cell-derived SEVs (HSEVs) exhibit increased therapeutic potency. We thus explored the role of HSEVs in macrophage immune regulation after spinal cord injury in rats and their significance in spinal cord repair. SEVs or HSEVs were isolated from bone marrow MSC supernatants by density gradient ultracentrifugation. HSEV administration to rats via tail vein injection after spinal cord injury reduced the lesion area and attenuated spinal cord inflammation. HSEVs regulate macrophage polarization towards the M2 phenotype in vivo and in vitro. MicroRNA sequencing and bioinformatics analyses of SEVs and HSEVs revealed that miR-146a-5p is a potent mediator of macrophage polarization that targets interleukin-1 receptor-associated kinase 1. Reducing miR-146a-5p expression in HSEVs partially attenuated macrophage polarization. Our data suggest that HSEVs attenuate spinal cord inflammation and injury in rats by transporting miR-146a-5p, which alters macrophage polarization. This study provides new insights into the application of HSEVs as a therapeutic tool for spinal cord injury.