Merck
  • Developing a human iPSC-derived three-dimensional myelin spheroid platform for modeling myelin diseases.
Developing a human iPSC-derived three-dimensional myelin spheroid platform for modeling myelin diseases.
iScience (2023-10-23)
Lizhao Feng, Jianfei Chao, Mingzi Zhang, Elizabeth Pacquing, Weidong Hu, Yanhong Shi
ABSTRACT
Myelin defects cause a collection of myelin disorders in the brain. The lack of human models has limited us from better understanding pathological mechanisms of myelin diseases. While human induced pluripotent stem cell (hiPSC)-derived spheroids or organoids have been used to study brain development and disorders, it has been difficult to recapitulate mature myelination in these structures. Here, we have developed a method to generate three-dimensional (3D) myelin spheroids from hiPSCs in a robust and reproducible manner. Using this method, we generated myelin spheroids from patient iPSCs to model Canavan disease (CD), a demyelinating disorder. By using CD patient iPSC-derived myelin spheroids treated with N-acetyl-aspartate (NAA), we were able to recapitulate key pathological features of the disease and show that high-level NAA is sufficient to induce toxicity on myelin sheaths. Our study has established a 3D human cellular platform to model human myelin diseases for mechanistic studies and drug discovery.
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAG Antibody, clone 513, Alexa Fluor 488 Conjugate, clone 513, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-APC Antibody, clone CC-1, clone CC-1, from mouse
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Caspr Antibody, clone K65/35, clone K65/35, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
SAG dihydrochloride, ≥98% (HPLC)