Merck
  • Hypoxia activates SREBP2 through Golgi disassembly in bone marrow-derived monocytes for enhanced tumor growth.
Hypoxia activates SREBP2 through Golgi disassembly in bone marrow-derived monocytes for enhanced tumor growth.
The EMBO journal (2023-10-02)
Ryuichi Nakahara, Sho Aki, Maki Sugaya, Haruka Hirose, Miki Kato, Keisuke Maeda, Daichi M Sakamoto, Yasuhiro Kojima, Miyuki Nishida, Ritsuko Ando, Masashi Muramatsu, Melvin Pan, Rika Tsuchida, Yoshihiro Matsumura, Hideyuki Yanai, Hiroshi Takano, Ryoji Yao, Shinsuke Sando, Masabumi Shibuya, Juro Sakai, Tatsuhiko Kodama, Hiroyasu Kidoya, Teppei Shimamura, Tsuyoshi Osawa
ABSTRACT
Bone marrow-derived cells (BMDCs) infiltrate hypoxic tumors at a pre-angiogenic state and differentiate into mature macrophages, thereby inducing pro-tumorigenic immunity. A critical factor regulating this differentiation is activation of SREBP2-a well-known transcription factor participating in tumorigenesis progression-through unknown cellular mechanisms. Here, we show that hypoxia-induced Golgi disassembly and Golgi-ER fusion in monocytic myeloid cells result in nuclear translocation and activation of SREBP2 in a SCAP-independent manner. Notably, hypoxia-induced SREBP2 activation was only observed in an immature lineage of bone marrow-derived cells. Single-cell RNA-seq analysis revealed that SREBP2-mediated cholesterol biosynthesis was upregulated in HSCs and monocytes but not in macrophages in the hypoxic bone marrow niche. Moreover, inhibition of cholesterol biosynthesis impaired tumor growth through suppression of pro-tumorigenic immunity and angiogenesis. Thus, our findings indicate that Golgi-ER fusion regulates SREBP2-mediated metabolic alteration in lineage-specific BMDCs under hypoxia for tumor progression.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PECAM-1 Antibody, clone 2H8, Azide Free, clone 2H8, Chemicon®, from hamster(Armenian)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial