Merck
  • Impaired islet function and normal exocrine enzyme secretion occur with low inter-regional variation in type 1 diabetes.
Impaired islet function and normal exocrine enzyme secretion occur with low inter-regional variation in type 1 diabetes.
Cell reports (2024-06-08)
Denise M Drotar, Ana Karen Mojica-Avila, Drew T Bloss, Christian M Cohrs, Cameron T Manson, Amanda L Posgai, MacKenzie D Williams, Maigan A Brusko, Edward A Phelps, Clive H Wasserfall, Stephan Speier, Mark A Atkinson
ABSTRACT
Histopathological heterogeneity in the human pancreas is well documented; however, functional evidence at the tissue level is scarce. Herein, we investigate in situ glucose-stimulated islet and carbachol-stimulated acinar cell secretion across the pancreas head (PH), body (PB), and tail (PT) regions in donors without diabetes (ND; n = 15), positive for one islet autoantibody (1AAb+; n = 7), and with type 1 diabetes (T1D; <14 months duration, n = 5). Insulin, glucagon, pancreatic amylase, lipase, and trypsinogen secretion along with 3D tissue morphometrical features are comparable across regions in ND. In T1D, insulin secretion and beta-cell volume are significantly reduced within all regions, while glucagon and enzymes are unaltered. Beta-cell volume is lower despite normal insulin secretion in 1AAb+, resulting in increased volume-adjusted insulin secretion versus ND. Islet and acinar cell secretion in 1AAb+ are consistent across the PH, PB, and PT. This study supports low inter-regional variation in pancreas slice function and, potentially, increased metabolic demand in 1AAb+.
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture