Merck
  • Identification of IQCH as a calmodulin-associated protein required for sperm motility in humans.
Identification of IQCH as a calmodulin-associated protein required for sperm motility in humans.
iScience (2023-07-31)
Emma Cavarocchi, Camille Sayou, Patrick Lorès, Caroline Cazin, Laurence Stouvenel, Elma El Khouri, Charles Coutton, Zine-Eddine Kherraf, Catherine Patrat, Jérôme Govin, Nicolas Thierry-Mieg, Marjorie Whitfield, Pierre F Ray, Emmanuel Dulioust, Aminata Touré
ABSTRACT
Sperm fertilization ability mainly relies on proper sperm progression through the female genital tract and capacitation, which involves phosphorylation signaling pathways triggered by calcium and bicarbonate. We performed exome sequencing of an infertile asthenozoospermic patient and identified truncating variants in MAP7D3, encoding a microtubule-associated protein, and IQCH, encoding a protein of unknown function with enzymatic and signaling features. We demonstrate the deleterious impact of both variants on sperm transcripts and proteins from the patient. We show that, in vitro, patient spermatozoa could not induce the phosphorylation cascades associated with capacitation. We also provide evidence for IQCH association with calmodulin, a well-established calcium-binding protein that regulates the calmodulin kinase. Notably, we describe IQCH spatial distribution around the sperm axoneme, supporting its function within flagella. Overall, our work highlights the cumulative pathological impact of gene mutations and identifies IQCH as a key protein required for sperm motility and capacitation.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial