Merck
  • MATR3 P154S knock-in mice do not exhibit motor, muscle or neuropathologic features of ALS.
MATR3 P154S knock-in mice do not exhibit motor, muscle or neuropathologic features of ALS.
Biochemical and biophysical research communications (2023-01-24)
Marissa Dominick, Nicole Houchins, Vinisha Venugopal, Aamir R Zuberi, Cathleen M Lutz, Bessie Meechooveet, Kendall Van Keuren-Jensen, Robert Bowser, David X Medina
ABSTRACT
Matrin 3 is a nuclear matrix protein that has many roles in RNA processing including splicing and transport of mRNA. Many missense mutations in the Matrin 3 gene (MATR3) have been linked to familial forms of amyotrophic lateral sclerosis (ALS) and distal myopathy. However, the exact role of MATR3 mutations in ALS and myopathy pathogenesis is not understood. To demonstrate a role of MATR3 mutations in vivo, we generated a novel CRISPR/Cas9 mediated knock-in mouse model harboring the MATR3 P154S mutation expressed under the control of the endogenous promoter. The P154S variant of the MATR3 gene has been linked to familial forms of ALS. Heterozygous and homozygous MATR3 P154S knock-in mice did not develop progressive motor deficits compared to wild-type mice. In addition, ALS-like pathology did not develop in nervous or muscle tissue in either heterozygous or homozygous mice. Our results suggest that the MATR3 P154S variant is not sufficient to produce ALS-like pathology in vivo.
Sigma-Aldrich
Anti-Neurofilament L Antibody, serum, Chemicon®