Merck
  • Myofiber Baf60c controls muscle regeneration by modulating Dkk3-mediated paracrine signaling.
Myofiber Baf60c controls muscle regeneration by modulating Dkk3-mediated paracrine signaling.
The Journal of experimental medicine (2023-06-07)
Jingya Xu, Xiaofei Li, Wei Chen, Ziyin Zhang, Yanping Zhou, Yahui Gou, Cheng-An Lv, Lu Jin, Xinyuan Qiu, Shengshan Ma, Qing-Qian Wu, Tongyu Liu, Lin Mi, Zhuoying Yang, Ting Yu, Xiaowen Pan, Yu Feng, Pengfei Shan, Zhuo-Xian Meng
ABSTRACT
Obesity and type 2 diabetes (T2D) are the leading causes of the progressive decline in muscle regeneration and fitness in adults. The muscle microenvironment is known to play a key role in controlling muscle stem cell regenerative capacity, yet the underlying mechanism remains elusive. Here, we found that Baf60c expression in skeletal muscle is significantly downregulated in obese and T2D mice and humans. Myofiber-specific ablation of Baf60c in mice impairs muscle regeneration and contraction, accompanied by a robust upregulation of Dkk3, a muscle-enriched secreted protein. Dkk3 inhibits muscle stem cell differentiation and attenuates muscle regeneration in vivo. Conversely, Dkk3 blockade by myofiber-specific Baf60c transgene promotes muscle regeneration and contraction. Baf60c interacts with Six4 to synergistically suppress myocyte Dkk3 expression. While muscle expression and circulation levels of Dkk3 are markedly elevated in obese mice and humans, Dkk3 knockdown improves muscle regeneration in obese mice. This work defines Baf60c in myofiber as a critical regulator of muscle regeneration through Dkk3-mediated paracrine signaling.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution