Merck
  • Tetherin antagonism by SARS-CoV-2 ORF3a and spike protein enhances virus release.
Tetherin antagonism by SARS-CoV-2 ORF3a and spike protein enhances virus release.
EMBO reports (2023-10-11)
Hazel Stewart, Roberta Palmulli, Kristoffer H Johansen, Naomi McGovern, Ola M Shehata, George W Carnell, Hannah K Jackson, Jin S Lee, Jonathan C Brown, Thomas Burgoyne, Jonathan L Heeney, Klaus Okkenhaug, Andrew E Firth, Andrew A Peden, James R Edgar
ABSTRACT
The antiviral restriction factor, tetherin, blocks the release of several different families of enveloped viruses, including the Coronaviridae. Tetherin is an interferon-induced protein that forms parallel homodimers between the host cell and viral particles, linking viruses to the surface of infected cells and inhibiting their release. We demonstrate that SARS-CoV-2 infection causes tetherin downregulation and that tetherin depletion from cells enhances SARS-CoV-2 viral titres. We investigate the potential viral proteins involved in abrogating tetherin function and find that SARS-CoV-2 ORF3a reduces tetherin localisation within biosynthetic organelles where Coronaviruses bud, and increases tetherin localisation to late endocytic organelles via reduced retrograde recycling. We also find that expression of Spike protein causes a reduction in cellular tetherin levels. Our results confirm that tetherin acts as a host restriction factor for SARS-CoV-2 and highlight the multiple distinct mechanisms by which SARS-CoV-2 subverts tetherin function.
Sigma-Aldrich
Anti-Cathepsin D Rabbit pAb, liquid, Calbiochem®
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial