Merck
  • Membrane binding of endocytic myosin-1s is inhibited by a class of ankyrin repeat proteins.
Membrane binding of endocytic myosin-1s is inhibited by a class of ankyrin repeat proteins.
Molecular biology of the cell (2023-08-02)
Alaina H Willet, Jun-Song Chen, Liping Ren, Kathleen L Gould
ABSTRACT
Myosin-1s are monomeric actin-based motors that function at membranes. Myo1 is the single myosin-1 isoform in Schizosaccharomyces pombe that works redundantly with Wsp1-Vrp1 to activate the Arp2/3 complex for endocytosis. Here, we identified Ank1 as an uncharacterized cytoplasmic Myo1 binding partner. We found that in ank1Δ cells, Myo1 dramatically redistributed from endocytic patches to decorate the entire plasma membrane and endocytosis was defective. Biochemical analysis and structural predictions suggested that the Ank1 ankyrin repeats bind the Myo1 lever arm and the Ank1 acidic tail binds the Myo1 TH1 domain to prevent TH1-dependent Myo1 membrane binding. Indeed, Ank1 overexpression precluded Myo1 membrane localization and recombinant Ank1 reduced purified Myo1 liposome binding in vitro. Based on biochemical and cell biological analyses, we propose budding yeast Ank1 and human OSTF1 are functional Ank1 orthologs and that cytoplasmic sequestration by small ankyrin repeat proteins is a conserved mechanism regulating myosin-1s in endocytosis.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2