Merck
CD8+ T cell targeting of tumor antigens presented by HLA-E.
Science advances (2024-05-10)
Ravi F Iyer, Marieke C Verweij, Sujit S Nair, David Morrow, Mandana Mansouri, Dimple Chakravarty, Teresa Beechwood, Christine Meyer, Luke Uebelhoer, Elvin J Lauron, Andrea Selseth, Nessy John, Tin Htwe Thin, Siarhei Dzedzik, Colin Havenar-Daughton, Michael K Axthelm, Janet Douglas, Alan Korman, Nina Bhardwaj, Ashutosh K Tewari, Scott Hansen, Daniel Malouli, Louis J Picker, Klaus Früh
ABSTRACT
The nonpolymorphic major histocompatibility complex E (MHC-E) molecule is up-regulated on many cancer cells, thus contributing to immune evasion by engaging inhibitory NKG2A/CD94 receptors on NK cells and tumor-infiltrating T cells. To investigate whether MHC-E expression by cancer cells can be targeted for MHC-E-restricted T cell control, we immunized rhesus macaques (RM) with rhesus cytomegalovirus (RhCMV) vectors genetically programmed to elicit MHC-E-restricted CD8+ T cells and to express established tumor-associated antigens (TAAs) including prostatic acidic phosphatase (PAP), Wilms tumor-1 protein, or Mesothelin. T cell responses to all three tumor antigens were comparable to viral antigen-specific responses with respect to frequency, duration, phenotype, epitope density, and MHC restriction. Thus, CMV-vectored cancer vaccines can bypass central tolerance by eliciting T cells to noncanonical epitopes. We further demonstrate that PAP-specific, MHC-E-restricted CD8+ T cells from RhCMV/PAP-immunized RM respond to PAP-expressing HLA-E+ prostate cancer cells, suggesting that the HLA-E/NKG2A immune checkpoint can be exploited for CD8+ T cell-based immunotherapies.
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)