Merck
  • 2-(4-Methylthiazol-5-yl) Ethyl Nitrate Hydrochloride Ameliorates Cognitive Impairment via Modulation of Oxidative Stress and Nuclear Factor Kappa B (NF-κB) Signaling Pathway in Chronic Cerebral Hypoperfusion-Associated Spontaneously Hypertensive Rats.
2-(4-Methylthiazol-5-yl) Ethyl Nitrate Hydrochloride Ameliorates Cognitive Impairment via Modulation of Oxidative Stress and Nuclear Factor Kappa B (NF-κB) Signaling Pathway in Chronic Cerebral Hypoperfusion-Associated Spontaneously Hypertensive Rats.
Antioxidants (Basel, Switzerland) (2024-05-25)
Jiang Li, Shaofeng Xu, Ling Wang, Xiaoliang Wang
ABSTRACT
Hypertension reduces the bioavailability of vascular nitric oxide (NO) and contributes to the onset of vascular dementia (VaD). A loss of NO bioavailability increases inflammation and oxidative stress. 2-(4-Methylthiazol-5-yl) ethyl nitrate hydrochloride (W1302) is a novel nitric oxide donor (NOD) which is undergoing phase I clinical trials in China for the treatment of VaD. In this study, we investigated the protective effects of W1302 in VaD rats induced by the permanent occlusion of a bilateral common carotid arteries model related to spontaneous hypertension (SHR-2VO), and we further explored the underlying mechanisms. Nimodipine was used as a positive control. Our results showed that W1302 treatment for 4 weeks (10 mg/Kg/day) exhibited stronger improvement in the spatial learning and memory deficits in SHR-2VO rats compared with nimodipine with slightly lower systolic blood pressure (SBP). Meanwhile, W1302 treatment significantly increased NO and cGMP production, restored mitochondrial membrane potential and attenuated oxidative stress as evidenced by increasing ATP production and reducing malondialdehyde (MDA) levels in the brain. Furthermore, W1302 treatment markedly inhibited the iNOS activity and decreased TNF-α expression via inhibiting the nuclear factor kappa B (NF-κB) signaling pathway. Nimodipine treatment also restored these aberrant changes, but its ATP production was weaker than that of W1302, and there was no significant effect on NO release. Taken together, W1302 exhibited beneficial effects on complications in VaD with hypertension, which is involved in suppressing oxidative damage, and the inflammatory reaction might be mediated by an increase in NO release. Therefore, W1302 has therapeutic potential for the treatment of VaD caused by chronic cerebral hypoperfusion-associated spontaneous hypertension.
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder