Merck
  • Association of Mitochondrial Pyruvate Carrier with the Clinical and Histological Features in Lupus Nephritis.
Association of Mitochondrial Pyruvate Carrier with the Clinical and Histological Features in Lupus Nephritis.
International journal of nephrology and renovascular disease (2024-01-24)
Huanhuan Zhu, Chen Chen, Luhan Geng, Qing Li, Chengning Zhang, Lin Wu, Bo Zhang, Suyan Duan, Changying Xing, Yanggang Yuan
ABSTRACT
Mounting evidence suggests that mitochondrial dysfunction contributes to lupus nephritis (LN) pathogenesis. Mitochondrial pyruvate carrier 1 (MPC1) and mitochondrial pyruvate carrier 2 (MPC2) mediating pyruvate transport from the cytoplasm to the mitochondrial matrix, determines the cell survival and cellular energy supply. Here, we aimed to investigate the association of mitochondrial pyruvate carrier expression with the clinical and histological features in LN. Patients with biopsy-proven proliferative LN (class III and class IV, n=18) and membranous LN (class V, n=18) were included. Expression of MPC1 and MPC2 were examined by immunohistochemistry. MPC protein levels in the two groups were evaluated by the Student's t-test. Correlation analysis between MPC levels and clinicopathological features was performed by Spearman's rank correlation. Both MPC1 and MPC2 were exclusively expressed in renal tubules of enrolled LN. Significantly lower MPC1 and MPC2 were observed in patients with proliferative LN compared to membranous LN. In addition, the MPC1 and MPC2 were negatively correlated with SLEDAI-2K score, renal function, and renal pathology activity index. Both MPC1 and MPC2 were localized in renal tubules, and decreased MPC content was more pronounced in proliferative LN than membranous LN. MPC levels were significantly correlated with renal functions and renal pathology activity.