Merck
  • Simultaneous extraction and clean-up of polychlorinated biphenyls and their metabolites from small tissue samples using pressurized liquid extraction.
Simultaneous extraction and clean-up of polychlorinated biphenyls and their metabolites from small tissue samples using pressurized liquid extraction.
Journal of chromatography. A (2008-11-21)
Izabela Kania-Korwel, Hongxia Zhao, Karin Norstrom, Xueshu Li, Keri C Hornbuckle, Hans-Joachim Lehmler
ABSTRACT
A pressurized liquid extraction-based method for the simultaneous extraction and in situ clean-up of polychlorinated biphenyls (PCBs), hydroxylated (OH)-PCBs and methylsulfonyl (MeSO(2))-PCBs from small (<0.5 g) tissue samples was developed and validated. Extraction of a laboratory reference material with hexane-dichloromethane-methanol (48:43:9, v/v) and Florisil as fat retainer allowed an efficient recovery of PCBs (78-112%; RSD: 13-37%), OH-PCBs (46+/-2%; RSD: 4%) and MeSO(2)-PCBs (89+/-21%; RSD: 24%). Comparable results were obtained with an established analysis method for PCBs, OH-PCBs and MeSO(2)-PCBs.
Product Description
Supelco
Aroclor 1242 solution, certified reference material, 50 mg/kg in transformer oil