Merck
  • Mechanism and regulation of cargo entry into the Commander endosomal recycling pathway.
Mechanism and regulation of cargo entry into the Commander endosomal recycling pathway.
Nature communications (2024-08-22)
Rebeka Butkovič, Alexander P Walker, Michael D Healy, Kerrie E McNally, Meihan Liu, Tineke Veenendaal, Kohji Kato, Nalan Liv, Judith Klumperman, Brett M Collins, Peter J Cullen
ABSTRACT
Commander is a multiprotein complex that orchestrates endosomal recycling of integral cargo proteins and is essential for normal development. While the structure of this complex has recently been described, how cargo proteins are selected for Commander-mediated recycling remains unclear. Here we identify the mechanism through which the unstructured carboxy-terminal tail of the cargo adaptor sorting nexin-17 (SNX17) directly binds to the Retriever sub-complex of Commander. SNX17 adopts an autoinhibited conformation where its carboxy-terminal tail occupies the cargo binding groove. Competitive cargo binding overcomes this autoinhibition, promoting SNX17 endosomal residency and the release of the tail for Retriever association. Furthermore, our study establishes the central importance of SNX17-Retriever association in the handover of integrin and lipoprotein receptor cargoes into pre-existing endosomal retrieval sub-domains. In describing the principal mechanism of cargo entry into the Commander recycling pathway we provide key insight into the function and regulation of this evolutionary conserved sorting pathway.
Sigma-Aldrich
Anti-DSCR3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture