Merck
  • NF-κB-Inducing Kinase Governs the Mitochondrial Respiratory Capacity, Differentiation, and Inflammatory Status of Innate Immune Cells.
NF-κB-Inducing Kinase Governs the Mitochondrial Respiratory Capacity, Differentiation, and Inflammatory Status of Innate Immune Cells.
Journal of immunology (Baltimore, Md. : 1950) (2023-03-08)
Justin N Keeney, Ashley D Winters, Raquel Sitcheran, A Phillip West
ABSTRACT
NF-κB-inducing kinase (NIK), which is essential for the activation of the noncanonical NF-κB pathway, regulates diverse processes in immunity, development, and disease. Although recent studies have elucidated important functions of NIK in adaptive immune cells and cancer cell metabolism, the role of NIK in metabolic-driven inflammatory responses in innate immune cells remains unclear. In this study, we demonstrate that murine NIK-deficient bone marrow-derived macrophages exhibit defects in mitochondrial-dependent metabolism and oxidative phosphorylation, which impair the acquisition of a prorepair, anti-inflammatory phenotype. Subsequently, NIK-deficient mice exhibit skewing of myeloid cells characterized by aberrant eosinophil, monocyte, and macrophage cell populations in the blood, bone marrow, and adipose tissue. Furthermore, NIK-deficient blood monocytes display hyperresponsiveness to bacterial LPS and elevated TNF-α production ex vivo. These findings suggest that NIK governs metabolic rewiring, which is critical for balancing proinflammatory and anti-inflammatory myeloid immune cell function. Overall, our work highlights a previously unrecognized role for NIK as a molecular rheostat that fine-tunes immunometabolism in innate immunity, and suggests that metabolic dysfunction may be an important driver of inflammatory diseases caused by aberrant NIK expression or activity.
Product Description
B022, ≥97% (HPLC)
Sigma-Aldrich
Anti-Tom20/Tomm20 Antibody, clone 2F8.1, clone 2F8.1, from mouse
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Anti-IKKα Mouse mAb (14A231), liquid, clone 14A231, Calbiochem®
Sigma-Aldrich
Anti-DNA Antibody, clone AC-30-10, clone AC-30-10, Chemicon®, from mouse