- Identification of a Biomarker Combination for Survival Stratification in pStage II/III Gastric Cancer after Curative Resection.
Identification of a Biomarker Combination for Survival Stratification in pStage II/III Gastric Cancer after Curative Resection.
We sought to identify an optimal combination of survival risk stratification markers in patients with pathological (p) stage II/III gastric cancer (GC) after curative resection. We measured the expression levels of 127 genes in pStage II/III GC tissues of two patient cohorts by quantitative polymerase chain reaction (qPCR) and the expression of 1756 proteins between two prognosis (good and poor) groups by proteomic analysis to identify candidate survival stratification markers. Further, immunohistochemistry (IHC) using tumor microarrays (TMAs) in another cohort of patients was performed to identify an optimal biomarker combination for survival stratification in GC patients. secreted protein acidic and rich in cysteine (SPARC), erb-b2 receptor tyrosine kinase 2 (ERBB2), inhibin subunit beta A (INHBA), matrix metallopeptidase-11 (MMP11), tumor protein p53 (TP53), and platelet-derived growth factor receptor-beta (PDGFRB) were identified as candidate biomarkers from qPCR analysis, and SPARC and galectin-10 were obtained from the proteomic analysis. The combination of PDGFRB, INHBA, MMP11, and galectin-10 was identified as the optimal combination of survival risk stratification markers. A combination of four proteins in GC tissues may serve as useful survival risk stratification markers in patients with pStage II/III GC following curative resection. Our results may facilitate future multicenter prospective clinical trials.