Merck
  • CD19-targeting CAR T cells protect from ANCA-induced acute kidney injury.
CD19-targeting CAR T cells protect from ANCA-induced acute kidney injury.
Annals of the rheumatic diseases (2024-01-06)
Dörte Lodka, Maria Zschummel, Mario Bunse, Anthony Rousselle, Janis Sonnemann, Ralph Kettritz, Uta E Höpken, Adrian Schreiber
ABSTRACT
Anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitides (AAV) are life-threatening systemic autoimmune diseases manifesting in the kidneys as necrotizing crescentic glomerulonephritis (NCGN). ANCA antigens are myeloperoxidase (MPO) or proteinase 3. Current treatments include steroids, cytotoxic drugs and B cell-depleting antibodies. The use of chimeric antigen receptor (CAR) T cells in autoimmune diseases is a promising new therapeutic approach. We tested the hypothesis that CAR T cells targeting CD19 deplete B cells, including MPO-ANCA-producing B cells, thereby protecting from ANCA-induced NCGN. We tested this hypothesis in a preclinical MPO-AAV mouse model. NCGN was established by immunisation of MPO-/- mice with murine MPO, followed by irradiation and transplantation with haematopoietic cells from wild-type mice alone or together with either CD19-targeting CAR T cells or control CAR T cells. CD19 CAR T cells efficiently migrated to and persisted in bone marrow, spleen, peripheral blood and kidneys for up to 8 weeks. CD19 CAR T cells, but not control CAR T cells, depleted B cells and plasmablasts, enhanced the MPO-ANCA decline, and most importantly protected from NCGN. Our proof-of-principle study may encourage further exploration of CAR T cells as a treatment for ANCA-vasculitis patients with the goal of drug-free remission.