Merck
Genetic influence on vascular smooth muscle cell apoptosis.
Cell death & disease (2024-06-09)
David G McVey, Catherine Andreadi, Peng Gong, Paulina J Stanczyk, Charles U Solomon, Lenka Turner, Liu Yan, Runji Chen, Junjun Cao, Christopher P Nelson, John R Thompson, Haojie Yu, Tom R Webb, Nilesh J Samani, Shu Ye
ABSTRACT
Vascular smooth muscle cell (VSMC) proliferation, migration, and apoptosis play important roles in many physiological processes and pathological conditions. To identify genetic influences on VSMC behavior, we measured these traits and undertook genome-wide association studies in primary umbilical artery-derived VSMCs from >2000 individuals. Although there were no genome-wide significant associations for VSMC proliferation or migration, genetic variants at two genomic loci (7p15.3 and 7q32.3) showed highly significant associations with VSMC apoptosis (P = 1.95 × 10-13 and P = 7.47 × 10-9, respectively). The lead variant at the 7p51.3 locus was associated with increased expression of the GSDME and PALS2 genes in VSMCs. Knockdown of GSDME or PALS2 in VSMCs attenuated apoptotic cell death. A protein co-immunoprecipitation assay indicated that GSDME complexed with PALS2. PALS2 knockdown attenuated activated caspase-3 and GSDME fragmentation, whilst GSDME knockdown also reduced activated caspase-3. These findings provide new insights into the genetic regulation of VSMC apoptosis, with potential utility for therapeutic development.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture