Merck
  • Antioxidant and Anti-Apoptotic Neuroprotective Effects of Cinnamon in Imiquimod-Induced Lupus.
Antioxidant and Anti-Apoptotic Neuroprotective Effects of Cinnamon in Imiquimod-Induced Lupus.
Antioxidants (Basel, Switzerland) (2024-07-27)
Georges Maalouly, Christine-Marie-Anne Martin, Yara Baz, Youakim Saliba, Anna-Maria Baramili, Nassim Fares
ABSTRACT
Despite accumulating evidence correlating oxidative stress with lupus disease activity, the brain redox pathways are still poorly investigated. Cinnamomum cassia, a widely used spice with powerful antioxidant properties, could be a novel therapeutic candidate in lupus. C57BL/6J female mice were divided into five groups: sham, sham-cinnamon, lupus, lupus-cinnamon starting from induction, and lupus-cinnamon starting two weeks before induction. Lupus was induced by skin application on the right ear with 1.25 mg of 5% imiquimod cream three times per week for six weeks. Cinnamomum cassia was given orally, five days per week, at 200 mg/kg. Concomitant to TLR7-MYD88 pathway activation, the p-NRF2/NRF2 and p-FOXO3/FOXO3 ratios were increased in the hippocampus and alleviated by cinnamon treatment. BCL-2 positivity was enhanced in hippocampal neurons and reversed only by preventive cinnamon administration. In vitro, exposure of hippocampal cells to the plasma of different groups induced a surge in oxidative stress. This was associated with an increased t-BID/BID ratio. Cinnamon treatment, particularly in the preventive arm, normalized these modifications. Our study shows a neuroprotective effect of cinnamon by rescuing brain redox and apoptosis homeostasis in lupus, paving the way for its use as a natural therapeutic compound in the clinical management of lupus.