Merck
  • Regulatory T cells use heparanase to access IL-2 bound to extracellular matrix in inflamed tissue.
Regulatory T cells use heparanase to access IL-2 bound to extracellular matrix in inflamed tissue.
Nature communications (2024-02-21)
Hunter A Martinez, Ievgen Koliesnik, Gernot Kaber, Jacqueline K Reid, Nadine Nagy, Graham Barlow, Ben A Falk, Carlos O Medina, Aviv Hargil, Svenja Zihsler, Israel Vlodavsky, Jin-Ping Li, Magdiel Pérez-Cruz, Sai-Wen Tang, Everett H Meyer, Lucile E Wrenshall, James D Lord, K Christopher Garcia, Theo D Palmer, Lawrence Steinman, Gerald T Nepom, Thomas N Wight, Paul L Bollyky, Hedwich F Kuipers
ABSTRACT
Although FOXP3+ regulatory T cells (Treg) depend on IL-2 produced by other cells for their survival and function, the levels of IL-2 in inflamed tissue are low, making it unclear how Treg access this critical resource. Here, we show that Treg use heparanase (HPSE) to access IL-2 sequestered by heparan sulfate (HS) within the extracellular matrix (ECM) of inflamed central nervous system tissue. HPSE expression distinguishes human and murine Treg from conventional T cells and is regulated by the availability of IL-2. HPSE-/- Treg have impaired stability and function in vivo, including in the experimental autoimmune encephalomyelitis (EAE) mouse model of multiple sclerosis. Conversely, endowing monoclonal antibody-directed chimeric antigen receptor (mAbCAR) Treg with HPSE enhances their ability to access HS-sequestered IL-2 and their ability to suppress neuroinflammation in vivo. Together, these data identify a role for HPSE and the ECM in immune tolerance, providing new avenues for improving Treg-based therapy of autoimmunity.