Merck
  • NlpI-Prc Proteolytic Complex Mediates Peptidoglycan Synthesis and Degradation via Regulation of Hydrolases and Synthases in Escherichia coli.
NlpI-Prc Proteolytic Complex Mediates Peptidoglycan Synthesis and Degradation via Regulation of Hydrolases and Synthases in Escherichia coli.
International journal of molecular sciences (2023-11-25)
Xinwei Liu, Tanneke den Blaauwen
ABSTRACT
Balancing peptidoglycan (PG) synthesis and degradation with precision is essential for bacterial growth, yet our comprehension of this intricate process remains limited. The NlpI-Prc proteolytic complex plays a crucial but poorly understood role in the regulation of multiple enzymes involved in PG metabolism. In this paper, through fluorescent D-amino acid 7-hydroxycoumarincarbonylamino-D-alanine (HADA) labeling and immunolabeling assays, we have demonstrated that the NlpI-Prc complex regulates the activity of PG transpeptidases and subcellular localization of PBP3 under certain growth conditions. PBP7 (a PG hydrolase) and MltD (a lytic transglycosylase) were confirmed to be negatively regulated by the NlpI-Prc complex by an in vivo degradation assay. The endopeptidases, MepS, MepM, and MepH, have consistently been demonstrated as redundantly essential "space makers" for nascent PG insertion. However, we observed that the absence of NlpI-Prc complex can alleviate the lethality of the mepS mepM mepH mutant. A function of PG lytic transglycosylases MltA and MltD as "space makers" was proposed through multiple gene deletions. These findings unveil novel roles for NlpI-Prc in the regulation of both PG synthesis and degradation, shedding light on the previously undiscovered function of lytic transglycosylases as "space makers" in PG expansion.
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution