Merck
  • Sulforaphane Effects on Neuronal-like Cells and Peripheral Blood Mononuclear Cells Exposed to 2.45 GHz Electromagnetic Radiation.
Sulforaphane Effects on Neuronal-like Cells and Peripheral Blood Mononuclear Cells Exposed to 2.45 GHz Electromagnetic Radiation.
International journal of molecular sciences (2024-07-27)
Maria Paola Bertuccio, Caterina Saija, Giuseppe Acri, Riccardo Ientile, Daniela Caccamo, Monica Currò
ABSTRACT
Exposure to 2.45 GHz electromagnetic radiation (EMR) emitted from commonly used devices has been reported to induce oxidative stress in several experimental models. Our study aims to evaluate the efficacy of sulforaphane, a well-known natural product, in preventing radiation-induced toxic effects caused by a 24 h exposure of SH-SY5Y neuronal-like cells and peripheral blood mononuclear cells (PBMCs) to 2.45 GHz EMR. Cells were exposed to radiation for 24 h in the presence or absence of sulforaphane at different concentrations (5-10-25 µg/mL). Cell viability, mitochondrial activity alterations, the transcription and protein levels of redox markers, and apoptosis-related genes were investigated. Our data showed a reduction in cell viability of both neuronal-like cells and PBMCs caused by EMR exposure and a protective effect of 5 µg/mL sulforaphane. The lowest sulforaphane concentration decreased ROS production and increased the Mitochondrial Transmembrane Potential (Δψm) and the NAD+/NADH ratio, which were altered by radiation exposure. Sulforaphane at higher concentrations displayed harmful effects. The hormetic behavior of sulforaphane was also evident after evaluating the expression of genes coding for Nrf2, SOD2, and changes in apoptosis markers. Our study underlined the vulnerability of neuronal-like cells to mitochondrial dysfunction and oxidative stress and the possibility of mitigating these effects by supplementation with sulforaphane. To our knowledge, there are no previous studies about the effects of SFN on these cells when exposed to 2.45 GHz electromagnetic radiation.