Merck
  • Expression and localization of apelin and apelin receptor protein in the oviduct of letrozole-induced hyperandrogenized mice.
Expression and localization of apelin and apelin receptor protein in the oviduct of letrozole-induced hyperandrogenized mice.
Cell biology international (2024-04-18)
Ayushmita Dutta, Borgohain Anima, Preethi Riba, Guruswami Gurusubramanian, Vikas Kumar Roy
ABSTRACT
Apelin and its receptor (APJ) are expressed in the reproductive organs of some mammalian females. The function of oviduct has also been suggested to be compromised in the hyperandrogenism condition. However, expression of apelin and APJ has not been shown in the oviduct of hyperandrogenized mice. Thus, the present study has investigated the localization and expression of apelin and APJ in the letrozole-induced hyperandrogenized mice oviduct. Histomorphometric analysis showed decreased lumen of oviduct in the hyperandrogenized mice. Our results showed elevated expression of APJ and decreased abundance of apelin in the hyperandrogenized mice oviduct. This finding suggests impaired apelin signaling in the oviduct of hyperandrogenized mice. The expression of androgen receptor was upregulated while estrogen receptors were downregulated in the hyperandrogenized mice. The expression of HSP70 was also downregulated along with increased expression of active caspase 3 and BAX and decreased expression of BCL2 in hyperandrogenized mice. Furthermore, the phosphorylation of phospho-Ser473-Akt and phospho-Thr308-Akt also showed differential levels in the oviduct of hyperandrogenized mice. Whether this differential phosphorylation of Akt was solely due to impaired apelin signaling in the oviduct, remains unclear. Moreover, increased androgen signaling and suppressed estrogen signaling coincides with elevated apoptosis. In conclusion, hyperandrogenized conditions could also impair the gamete transport and fertilization process due to apoptosis in the oviduct. However, further study would be required to unravel the exact role of apelin signaling in the oviduct in relation to apoptosis.
Sigma-Aldrich
Anti-Apelin receptor Antibody, from rabbit, purified by affinity chromatography