Merck
  • Impaired Mitochondrial Function and Marrow Failure in Patients Carrying a Variant of the SRSF4 Gene.
Impaired Mitochondrial Function and Marrow Failure in Patients Carrying a Variant of the SRSF4 Gene.
International journal of molecular sciences (2024-02-24)
Maurizio Miano, Nadia Bertola, Alice Grossi, Gianluca Dell'Orso, Stefano Regis, Marta Rusmini, Paolo Uva, Diego Vozzi, Francesca Fioredda, Elena Palmisani, Michela Lupia, Marina Lanciotti, Federica Grilli, Fabio Corsolini, Luca Arcuri, Maria Carla Giarratana, Isabella Ceccherini, Carlo Dufour, Enrico Cappelli, Silvia Ravera
ABSTRACT
Serine/arginine-rich splicing factors (SRSFs) are a family of proteins involved in RNA metabolism, including pre-mRNA constitutive and alternative splicing. The role of SRSF proteins in regulating mitochondrial activity has already been shown for SRSF6, but SRSF4 altered expression has never been reported as a cause of bone marrow failure. An 8-year-old patient admitted to the hematology unit because of leukopenia, lymphopenia, and neutropenia showed a missense variant of unknown significance of the SRSF4 gene (p.R235W) found via whole genome sequencing analysis and inherited from the mother who suffered from mild leuko-neutropenia. Both patients showed lower SRSF4 protein expression and altered mitochondrial function and energetic metabolism in primary lymphocytes and Epstein-Barr-virus (EBV)-immortalized lymphoblasts compared to healthy donor (HD) cells, which appeared associated with low mTOR phosphorylation and an imbalance in the proteins regulating mitochondrial biogenesis (i.e., CLUH) and dynamics (i.e., DRP1 and OPA1). Transfection with the wtSRSF4 gene restored mitochondrial function. In conclusion, this study shows that the described variant of the SRSF4 gene is pathogenetic and causes reduced SRSF4 protein expression, which leads to mitochondrial dysfunction. Since mitochondrial function is crucial for hematopoietic stem cell maintenance and some genetic bone marrow failure syndromes display mitochondrial defects, the SRSF4 mutation could have substantially contributed to the clinical phenotype of our patient.
Sigma-Aldrich
Antimycin A from Streptomyces sp.