Merck
  • Microglial ferroptotic stress causes non-cell autonomous neuronal death.
Microglial ferroptotic stress causes non-cell autonomous neuronal death.
Molecular neurodegeneration (2024-02-06)
Jeffrey R Liddell, James B W Hilton, Kai Kysenius, Jessica L Billings, Sara Nikseresht, Lachlan E McInnes, Dominic J Hare, Bence Paul, Stephen W Mercer, Abdel A Belaidi, Scott Ayton, Blaine R Roberts, Joseph S Beckman, Catriona A McLean, Anthony R White, Paul S Donnelly, Ashley I Bush, Peter J Crouch
ABSTRACT
Ferroptosis is a form of regulated cell death characterised by lipid peroxidation as the terminal endpoint and a requirement for iron. Although it protects against cancer and infection, ferroptosis is also implicated in causing neuronal death in degenerative diseases of the central nervous system (CNS). The precise role for ferroptosis in causing neuronal death is yet to be fully resolved. To elucidate the role of ferroptosis in neuronal death we utilised co-culture and conditioned medium transfer experiments involving microglia, astrocytes and neurones. We ratified clinical significance of our cell culture findings via assessment of human CNS tissue from cases of the fatal, paralysing neurodegenerative condition of amyotrophic lateral sclerosis (ALS). We utilised the SOD1G37R mouse model of ALS and a CNS-permeant ferroptosis inhibitor to verify pharmacological significance in vivo. We found that sublethal ferroptotic stress selectively affecting microglia triggers an inflammatory cascade that results in non-cell autonomous neuronal death. Central to this cascade is the conversion of astrocytes to a neurotoxic state. We show that spinal cord tissue from human cases of ALS exhibits a signature of ferroptosis that encompasses atomic, molecular and biochemical features. Further, we show the molecular correlation between ferroptosis and neurotoxic astrocytes evident in human ALS-affected spinal cord is recapitulated in the SOD1G37R mouse model where treatment with a CNS-permeant ferroptosis inhibitor, CuII(atsm), ameliorated these markers and was neuroprotective. By showing that microglia responding to sublethal ferroptotic stress culminates in non-cell autonomous neuronal death, our results implicate microglial ferroptotic stress as a rectifiable cause of neuronal death in neurodegenerative disease. As ferroptosis is currently primarily regarded as an intrinsic cell death phenomenon, these results introduce an entirely new pathophysiological role for ferroptosis in disease.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat