Merck
  • Retinal Ganglion Cell Fate Induction by Ngn-Family Transcription Factors.
Retinal Ganglion Cell Fate Induction by Ngn-Family Transcription Factors.
Investigative ophthalmology & visual science (2023-12-22)
Ke Zhang, Wenwen Cai, Yanling Xin, Qinghai He, Canbin Chen, Mingbing Zeng, Shuyi Chen
ABSTRACT
Retinal ganglion cells (RGCs) are the projection neurons of the retina. Loss of RGCs is the cellular basis for vision loss in patients with glaucoma. Finding ways to regenerate RGCs will aid in the development of regenerative therapies for patients with glaucoma. The aim of this study was to examine the ability of Ngn-family transcription factors (TFs) to induce RGC regeneration through reprogramming in vitro and in vivo. In vitro, lentiviruses were used to deliver Ngn-TFs into mouse embryonic fibroblasts (MEFs). In vivo, mouse pup retina electroporation was used to deliver Ngn-TFs into late-stage retinal progenitor cells (RPCs). Immunofluorescence staining and RNA sequencing were used to examine cell fate reprogramming; patch-clamp recording was used to examine neuronal electrophysiologic functions. In vitro, all three Ngn-TFs, Ngn1, Ngn2, and Ngn3, were able to work alone to reprogram MEFs into RGC-like neurons that resembled RGCs at the transcriptome level, exhibited typical neuronal membrane electrophysiologic properties, and formed functional synaptic communications with retinal neurons. In vivo, Ngn-TFs reprogrammed the differentiation-competent state of late-stage RPCs to generate RGCs. Ngn-TFs are effective in inducing an RGC-like fate both in vitro and in vivo and might be explored further in the future for glaucoma translational applications.
Sigma-Aldrich
Anti-Brn-3a Antibody, POU-domain protein, clone 5A3.2, culture supernatant, clone 5A3.2, Chemicon®
Sigma-Aldrich
Anti-Chx10 Antibody, CT, Chemicon®, from sheep
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®