Merck
  • Sex differences in basal reelin levels in the paraventricular hypothalamus and in response to chronic stress induced by repeated corticosterone in rats.
Sex differences in basal reelin levels in the paraventricular hypothalamus and in response to chronic stress induced by repeated corticosterone in rats.
Hormones and behavior (2022-10-25)
Carla L Sánchez-Lafuente, Raquel Romay-Tallon, Josh Allen, Jenessa N Johnston, Lisa E Kalynchuk, Hector J Caruncho
ABSTRACT
Repeated exposure to the stress hormone corticosterone results in depressive-like behaviours paralleled by the downregulation of hippocampal reelin expression. Reelin is expressed in key neural populations involved in the stress response, but whether its hypothalamic expression is sex-specific or involved in sex-specific vulnerability to stress is unknown. Female and male rats were treated with either daily vehicle or corticosterone injections (40 mg/kg) for 21 days. Thereafter, they were subjected to several behavioural tasks before being sacrificed to allow the analysis of reelin expression in hypothalamic nuclei. The basal density of reelin-positive cells in males was significantly higher in the paraventricular nucleus (19 %) and in the medial preoptic area (51 %) compared to females. Chronic corticosterone injections increased the immobility time in the forced swim test in males (107 %) and females (108 %) and decreased the exploration of the elevated plus maze in males (34 %). Corticosterone also caused a significant decrease in the density of reelin-positive cells in males, in both ventrodorsal (37 %) and ventrolateral (32 %) subdivisions of the paraventricular nucleus, while not affecting females. Moreover, in the paraventricular nucleus of males, 30 % of the basal reelin-positive cells co-expressed oxytocin while only 17.5 % did in females, showing a positive correlation between reelin and oxytocin levels. Chronic corticosterone did not significantly affect co-localization levels. For the first time, this study shows that there is a sexually dimorphic subpopulation of reelin-positive neurons in the paraventricular nucleus that can be differentially affected by chronic stress.
Sigma-Aldrich
Anti-Oxytocin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse