Merck
  • High-resolution transcriptomic and epigenetic profiling identifies novel regulators of COPD.
High-resolution transcriptomic and epigenetic profiling identifies novel regulators of COPD.
The EMBO journal (2023-05-05)
Uwe Schwartz, Maria Llamazares Prada, Stephanie T Pohl, Mandy Richter, Raluca Tamas, Michael Schuler, Corinna Keller, Vedrana Mijosek, Thomas Muley, Marc A Schneider, Karsten Quast, Joschka Hey, Claus P Heußel, Arne Warth, Hauke Winter, Özdemirhan Serçin, Harry Karmouty-Quintana, Soma Sk Jyothula, Manish K Patel, Felix Herth, Ina Koch, Giuseppe Petrosino, Alexandru Titimeaua, Balca R Mardin, Dieter Weichenhan, Tomasz P Jurkowski, Charles D Imbusch, Benedikt Brors, Vladimir Benes, Birgit Jung, David Wyatt, Heiko F Stahl, Christoph Plass, Renata Z Jurkowska
ABSTRACT
Patients with chronic obstructive pulmonary disease (COPD) are still waiting for curative treatments. Considering its environmental cause, we hypothesized that COPD will be associated with altered epigenetic signaling in lung cells. We generated genome-wide DNA methylation maps at single CpG resolution of primary human lung fibroblasts (HLFs) across COPD stages. We show that the epigenetic landscape is changed early in COPD, with DNA methylation changes occurring predominantly in regulatory regions. RNA sequencing of matched fibroblasts demonstrated dysregulation of genes involved in proliferation, DNA repair, and extracellular matrix organization. Data integration identified 110 candidate regulators of disease phenotypes that were linked to fibroblast repair processes using phenotypic screens. Our study provides high-resolution multi-omic maps of HLFs across COPD stages. We reveal novel transcriptomic and epigenetic signatures associated with COPD onset and progression and identify new candidate regulators involved in the pathogenesis of chronic lung diseases. The presence of various epigenetic factors among the candidates demonstrates that epigenetic regulation in COPD is an exciting research field that holds promise for novel therapeutic avenues for patients.