Merck
  • Loss of Dja2 accompanies pH deviation in lysosomes and lysosome-related organelles.
Loss of Dja2 accompanies pH deviation in lysosomes and lysosome-related organelles.
Journal of cellular physiology (2023-12-18)
Kazutoyo Terada, Motoyoshi Endo, Hiroshi Kiyonari, Naoki Takeda, Yuichi Oike
ABSTRACT
The Dja2 knockout (Dja2-/- ) mice had respiratory distress, and >60% died within 2 days after birth. The surviving adult Dja2-/- mice were infertile and the lungs of Dja2-/- mice showed several abnormalities, including the processing defect of prosurfactant protein C in the alveolar epithelial type II cells and the accumulation of glycolipids in enlarged alveolar macrophages. The luminal pH of acidic organelles in Dja2-/- cells was shifted to pH 5.37-5.45. This deviated pH was immediately restored to control levels (pH 4.56-4.65) by the addition of a diuretic, ethyl isopropyl amiloride (EIPA). Although the role of DJA2 in maintaining the pH homeostasis of lysosome-related organelles is currently obscure, this rapid and remarkable pH resilience is best explained by an EIPA-sensitive proton efflux machinery that is disorganized and overactivated due to the loss of Dja2.
Sigma-Aldrich
Anti-Glycosaminoglycan Antibody, skin specific Antibody, clone PG-4, clone PG-4, from mouse