Merck
  • Viral delivery of L1CAM promotes axonal extensions by embryonic cerebral grafts in mouse brain.
Viral delivery of L1CAM promotes axonal extensions by embryonic cerebral grafts in mouse brain.
Stem cell reports (2023-03-25)
Ryosuke Tsuchimochi, Keitaro Yamagami, Naoko Kubo, Naoya Amimoto, Fabian Raudzus, Bumpei Samata, Tetsuhiro Kikuchi, Daisuke Doi, Koji Yoshimoto, Aya Mihara, Jun Takahashi
ABSTRACT
Cell replacement therapy is expected as a new and more radical treatment against brain damage. We previously reported that transplanted human cerebral organoids extend their axons along the corticospinal tract in rodent brains. The axons reached the spinal cord but were still sparse. Therefore, this study optimized the host brain environment by the adeno-associated virus (AAV)-mediated expression of axon guidance proteins in mouse brain. Among netrin-1, SEMA3, and L1CAM, only L1CAM significantly promoted the axonal extension of mouse embryonic brain tissue-derived grafts. L1CAM was also expressed by donor neurons, and this promotion was exerted in a haptotactic manner by their homophilic binding. Primary cortical neurons cocultured on L1CAM-expressing HEK-293 cells supported this mechanism. These results suggest that optimizing the host environment by the AAV-mediated expression of axon guidance molecules enhances the effect of cell replacement therapy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Neuropilin-1 Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Neuropilin-2 Antibody, serum, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)