Merck
  • High glucose impairs osteogenic differentiation of embryonic stem cells via early diversion of beta-catenin from Forkhead box O to T cell factor interaction.
High glucose impairs osteogenic differentiation of embryonic stem cells via early diversion of beta-catenin from Forkhead box O to T cell factor interaction.
Birth defects research (2022-09-28)
Anke Dienelt, Kevin C Keller, Nicole I Zur Nieden
ABSTRACT
Diabetes, which is characterized by an increase in blood glucose concentration, is accompanied by low bone turnover, increased fracture risk, and the formation of embryonic skeletal malformations. Yet, there are few studies elucidating the underlying alterations in signaling pathways leading to these osteogenic defects. We hypothesized here that bone formation deficiencies in a high glucose environment result from altered activity of beta-catenin (CTNNB1), a key contributor to osteogenic differentiation, dysregulation of which has also been implicated in the development of diabetes. To test this hypothesis, we used a previously established embryonic stem cell (ESC) model of differentiation that mimics the diabetic environment of the developing embryo. We differentiated murine ESCs within osteogenic-inducing media containing either high (diabetic) or low (physiological) levels of D-glucose and performed time course analyses to study the influence of high glucose on early and late bone cell differentiation. Endpoint measures for osteogenic differentiation were reduced in a glucose-dependent manner and expression of precursor-specific markers altered at multiple time points. Furthermore, transcriptional activity of the lymphoid enhancer factor (LEF)/T cell factor (TCF) transcription factors during precursor formation stages was significantly elevated while levels of CTNNB1 complexed with Forkhead box O 3a (FOXO3a) declined. Modulation of AKT, a known upstream regulator of both LEF/TCF and FOXO3a, as well as CTNNB1 rescued some of the reductions in osteogenic output seen in the high glucose condition. Within our in vitro model, we found a clear involvement of LEF/TCF and FOXO3a signaling pathways in the regulation of osteogenic differentiation, which may account for the skeletal deficiencies found in newborns of diabetic mothers.
Sigma-Aldrich
Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone 11E6, clone 11E6, Upstate®, from mouse
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, clone AW24, rabbit monoclonal, clone AW24, Upstate®, from rabbit