Merck
  • Inceptor correlates with markers of prostate cancer progression and modulates insulin/IGF1 signaling and cancer cell migration.
Inceptor correlates with markers of prostate cancer progression and modulates insulin/IGF1 signaling and cancer cell migration.
Molecular metabolism (2023-03-18)
Katharina Wissmiller, Sara Bilekova, Andras Franko, Stefan Z Lutz, Miriam Katsburg, Sebastian Gulde, Natalia S Pellegata, Arnulf Stenzl, Martin Heni, Lucia Berti, Hans-Ulrich Häring, Heiko Lickert
ABSTRACT
The insulin/insulin-like growth factor 1 (IGF1) pathway is emerging as a crucial component of prostate cancer progression. Therefore, we investigated the role of the novel insulin/IGF1 signaling modulator inceptor in prostate cancer. We analyzed the expression of inceptor in human samples of benign prostate epithelium and prostate cancer. Further, we performed signaling and functional assays using prostate cancer cell lines. We found that inceptor was expressed in human benign and malignant prostate tissue and its expression positively correlated with various genes of interest, including genes involved in androgen signaling. In vitro, total levels of inceptor were increased upon androgen deprivation and correlated with high levels of androgen receptor in the nucleus. Inceptor overexpression was associated with increased cell migration, altered IGF1R trafficking and higher IGF1R activation. Our in vitro results showed that inceptor expression was associated with androgen status, increased migration, and IGF1R signaling. In human samples, inceptor expression was significantly correlated with markers of prostate cancer progression. Taken together, these data provide a basis for investigation of inceptor in the context of prostate cancer.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)