Merck
  • RAD51AP1 regulates ALT-HDR through chromatin-directed homeostasis of TERRA.
RAD51AP1 regulates ALT-HDR through chromatin-directed homeostasis of TERRA.
Molecular cell (2022-10-21)
Nicole Kaminski, Anne R Wondisford, Youngho Kwon, Michelle Lee Lynskey, Ragini Bhargava, Jonathan Barroso-González, Laura García-Expósito, Boxue He, Meng Xu, Dattatreya Mellacheruvu, Simon C Watkins, Mauro Modesti, Kyle M Miller, Alexey I Nesvizhskii, Huaiying Zhang, Patrick Sung, Roderick J O'Sullivan
ABSTRACT
Alternative lengthening of telomeres (ALT) is a homology-directed repair (HDR) mechanism of telomere elongation that controls proliferation in subsets of aggressive cancer. Recent studies have revealed that telomere repeat-containing RNA (TERRA) promotes ALT-associated HDR (ALT-HDR). Here, we report that RAD51AP1, a crucial ALT factor, interacts with TERRA and utilizes it to generate D- and R-loop HR intermediates. We also show that RAD51AP1 binds to and might stabilize TERRA-containing R-loops as RAD51AP1 depletion reduces R-loop formation at telomere DNA breaks. Proteomic analyses uncover a role for RAD51AP1-mediated TERRA R-loop homeostasis in a mechanism of chromatin-directed suppression of TERRA and prevention of transcription-replication collisions (TRCs) during ALT-HDR. Intriguingly, we find that both TERRA binding and this non-canonical function of RAD51AP1 require its intrinsic SUMO-SIM regulatory axis. These findings provide insights into the multi-contextual functions of RAD51AP1 within the ALT mechanism and regulation of TERRA.
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Anti-Ubiquityl Histone H2A.X (Lys119) Antibody, serum, from rabbit
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
RO-3306, ≥98% (HPLC)