Merck
  • DCAF14 regulates CDT2 to promote SET8-dependent replication fork protection.
DCAF14 regulates CDT2 to promote SET8-dependent replication fork protection.
Life science alliance (2023-11-09)
Neysha Tirado-Class, Caitlin Hathaway, Anthony Nelligan, Thuan Nguyen, Huzefa Dungrawala
ABSTRACT
DDB1- and CUL4-associated factors (DCAFs) CDT2 and DCAF14 are substrate receptors for Cullin4-RING E3 ubiquitin ligase (CRL4) complexes. CDT2 is responsible for PCNA-coupled proteolysis of substrates CDT1, p21, and SET8 during S-phase of cell cycle. DCAF14 functions at stalled replication forks to promote genome stability, but the mechanism is unknown. We find that DCAF14 mediates replication fork protection by regulating CRL4CDT2 activity. Absence of DCAF14 causes increased proteasomal degradation of CDT2 substrates. When forks are challenged with replication stress, increased CDT2 function causes stalled fork collapse and impairs fork recovery in DCAF14-deficient conditions. We further show that stalled fork protection is dependent on CDT2 substrate SET8 and does not involve p21 and CDT1. Like DCAF14, SET8 blocks nuclease-mediated digestion of nascent DNA at remodeled replication forks. Thus, unregulated CDT2-mediated turnover of SET8 triggers nascent strand degradation when DCAF14 is absent. We propose that DCAF14 controls CDT2 activity at stalled replication forks to facilitate SET8 function in safeguarding genomic integrity.
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Sigma-Aldrich
Anti-CDT2 Antibody, serum, from rabbit
Sigma-Aldrich
Anti-SETD8 (hPR-SET7) Antibody, from rabbit, purified by affinity chromatography