Merck
  • RAB27B expression in pancreatic cancer is predictive of poor survival but good response to chemotherapy.
RAB27B expression in pancreatic cancer is predictive of poor survival but good response to chemotherapy.
Cancer biomarkers : section A of Disease markers (2023-05-30)
Mathieu Pecqueux, Beate Wende, Ulrich Sommer, Franziska Baenke, Florian Oehme, Sebastian Hempel, Daniela Aust, Marius Distler, Jürgen Weitz, Christoph Kahlert
ABSTRACT
Pancreatic cancer is the 4th leading cause of cancer-related death with poor survival even after curative resection. RAB27A and RAB27B are key players in the exosome pathway where they play important roles in exosome secretion. Evidence suggests that RAB27A and RAB27B expression not only leads to tumor proliferation and invasion, but also plays an important role in antigen transfer necessary for anticancer immunity. In this study, we analyze the expression of RAB27A and RAB27B in patients after pancreatic cancer surgery with or without adjuvant chemotherapy and its influence on overall survival. We analyzed a total of 167 patients with pancreatic cancer for their RAB27A and RAB27B expression. We dichotomized the patients along the median and compared survival in patients with high and low RAB27A and RAB27B expression with or without adjuvant chemotherapy treatment. We found a significant improvement in overall survival in patients with a negative resection margin (p= 0.037) and in patients who received adjuvant chemotherapy (p= 0.039). The survival benefit after chemotherapy was dependent on RAB27B expression status: only the subgroup of patients with high RAB27B expression benefited from adjuvant chemotherapy (p= 0.006), but not the subgroup with low RAB27B expression (p= 0.59). Patients with high RAB27B expression who did not receive adjuvant chemotherapy showed a trend towards worse survival compared to the other subgroups. This difference was abolished after treatment with adjuvant chemotherapy. These results suggest that RAB27B expression in pancreatic cancer might identify a subgroup of patients with poor survival who might respond well to adjuvant chemotherapy. If resectable, these patients could be considered for neoadjuvant chemotherapy to minimize the risk of not receiving adjuvant chemotherapy. Further prospective studies are needed to confirm these findings.