Merck
  • Peroxynitrite in the tumor microenvironment changes the profile of antigens allowing escape from cancer immunotherapy.
Peroxynitrite in the tumor microenvironment changes the profile of antigens allowing escape from cancer immunotherapy.
Cancer cell (2022-10-12)
Evgenii N Tcyganov, Emilio Sanseviero, Douglas Marvel, Thomas Beer, Hsin-Yao Tang, Peter Hembach, David W Speicher, Qianfei Zhang, Laxminarasimha R Donthireddy, Ali Mostafa, Sabina Tsyganova, Vladimir Pisarev, Terri Laufer, Dmitriy Ignatov, Soldano Ferrone, Christiane Meyer, Hélène Maby-El Hajjami, Daniel E Speiser, Sooner Altiok, Scott Antonia, Xiaowei Xu, Wei Xu, Cathy Zheng, Lynn M Schuchter, Ravi K Amaravadi, Tara C Mitchell, Giorgos C Karakousis, Zhe Yuan, Luis J Montaner, Esteban Celis, Dmitry I Gabrilovich
ABSTRACT
Cancer immunotherapy often depends on recognition of peptide epitopes by cytotoxic T lymphocytes (CTLs). The tumor microenvironment (TME) is enriched for peroxynitrite (PNT), a potent oxidant produced by infiltrating myeloid cells and some tumor cells. We demonstrate that PNT alters the profile of MHC class I bound peptides presented on tumor cells. Only CTLs specific for PNT-resistant peptides have a strong antitumor effect in vivo, whereas CTLs specific for PNT-sensitive peptides are not effective. Therapeutic targeting of PNT in mice reduces resistance of tumor cells to CTLs. Melanoma patients with low PNT activity in their tumors demonstrate a better clinical response to immunotherapy than patients with high PNT activity. Our data suggest that intratumoral PNT activity should be considered for the design of neoantigen-based therapy and also may be an important immunotherapeutic target.