Merck
  • Clinical and Histopathologic Characteristics of Pediatric Patients With Primary Membranous Nephropathy.
Clinical and Histopathologic Characteristics of Pediatric Patients With Primary Membranous Nephropathy.
Kidney international reports (2023-11-29)
Anne M Kouri, Tiffany N Caza, Laurence H Beck, Jason M Misurac, Michael D Evans, Carrie L Phillips, Michael T Eadon, Christopher P Larsen, Sharon P Andreoli, Lihong Bu, Michelle N Rheault, Myda Khalid
ABSTRACT
Primary membranous nephropathy (PMN) is uncommon in children. Therefore, data on the clinical course of affected children are scarce. In recent years, several novel antigens have been implicated in the pathogenesis of PMN. However, the histopathologic characteristics of pediatric patients with PMN remain poorly represented in the literature. We have retrospectively analyzed the clinical presentation and outcomes data of 21 children with PMN from 3 centers in the United States. In addition, we have identified novel antigens in biopsy specimens from these patients and correlated their presence or absence to clinical outcomes. Finally, we compared the results of the novel antigen staining from our clinical cohort to a validation cohort of 127 biopsy specimens from children with PMN at Arkana Laboratories. The data from the 2 cohorts demonstrated similar overall antigen positivity rates of 62% to 63%, with phospholipase A2 receptor (PLA2R) and exostosin 1 (EXT1) being the most commonly found antigens. Results from the clinical cohort showed that overall, the kidney prognosis for children with PMN was good, with 17 of 21 patients entering a complete or partial remission. Children who were positive for PLA2R or EXT1 were significantly more likely to enter remission than those in the antigen negative group. Approximately 60% of pediatric membranous cases are positive for a novel antigen on kidney biopsy and the clinical prognosis is generally favorable. More studies are needed to understand the clinical implications of each specific novel antigen.