Merck
  • Inhibition of astroglial hemichannels ameliorates infrasonic noise induced short-term learning and memory impairment.
Inhibition of astroglial hemichannels ameliorates infrasonic noise induced short-term learning and memory impairment.
Behavioral and brain functions : BBF (2023-12-19)
Wei Zhang, Jue Yin, Bei-Yao Gao, Xi Lu, Ya-Jing Duan, Xu-Yan Liu, Ming-Zhen Li, Shan Jiang
ABSTRACT
As a kind of environmental noise, infrasonic noise has negative effects on various human organs. To date, research has shown that infrasound impairs cognitive function, especially the ability for learning and memory. Previously, we demonstrated that impaired learning and memory induced by infrasound was closely related with glia activation; however, the underlying mechanisms remain unclear. Connexin 43 hemichannels (Cx43 HCs), which are mainly expressed in hippocampal astrocytes, are activated under pathological conditions, lending support to the hypothesis that Cx43 HCs might function in the impaired learning and memory induced by infrasound. This study revealed that that blocking hippocampal Cx43 HCs or downregulating hippocampal Cx43 expression significantly alleviated impaired learning and memory induced by infrasound. We also observed that infrasound exposure led to the abundant release of glutamate and ATP through Cx43 HCs. In addition, the abundant release of glutamate and ATP depended on proinflammatory cytokines. Our finds suggested that the enhanced release of ATP and glutamate by astroglial Cx43 HCs may be involved in the learning and memory deficits caused by infrasound exposure.
Sigma-Aldrich
TAT-Gap19 trifluoroacetate salt, ≥95% (HPLC)