Merck
  • Fibulin-2 Facilitates Malignant Progression of Hepatocellular Carcinoma.
Fibulin-2 Facilitates Malignant Progression of Hepatocellular Carcinoma.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology (2023-05-10)
Xinyan Hu, Tianze Liu, Luting Li, Hairun Gan, Tiancheng Wang, Pengfei Pang, Junjie Mao
ABSTRACT
Identification of biomarkers to assist in the clinical management of hepatocellular carcinoma represents an urgent requirement. Fibulin-2 is known to contribute to the development and progression of various cancer types. This research investigated the role of fibulin-2 in hepatocellular carcinoma and explored the possible mechanisms. The expression of fibulin-2 in hepatocellular carcinoma was measured by bioinformatic analysis and confirmed by western blot and immunohistochemical staining in cell lines or patients' samples. The clinicopathologic features of hepatocellular carcinoma patients was analyzed. Cell viability assays were used to explore the role of fibulin-2 on proliferation in hepatocellular carcinoma. Western blot was conducted to uncover changes of protein expression of Ras-MEK-ERK1/2 pathway when Fibulin-2 was overexpressed or silenced. Flow cytometry analyses were used to determine the roles of fibulin-2 in the function of apoptosis and cell cycle. Subcutaneous xenograft mouse models showed the tumor growth pattern after fibulin-2 silence in vivo. We reported the upregulation of fibulin-2 in most hepatocellular carcinoma tissues and cells lines. Fibulin-2 promoted the proliferation of hepatocellular carcinoma cells in vitro by regulating Ras-MEK-ERK1/2 signaling pathway, whereas knockdown of fibulin-2 incurred the opposite effect on proliferation. Consistently, knockdown of fibulin-2 resulted in increased apoptosis and induced growth arrest during the G0/G1 phase transition. In vivo xenograft assessment confirmed that knockdown of fibulin-2 inhibited hepatocellular carcinoma tumor growth. Fibulin-2 exhibited tumor promotor activities in malignant progression of hepatocellular carcinoma. The results of the study highlighted the potential of fibulin-2 to be utilized as a promising biomarker and therapeutic target for hepatocellular carcinoma.