Merck
  • Cytoskeletal association of ATP citrate lyase controls the mechanodynamics of macropinocytosis.
Cytoskeletal association of ATP citrate lyase controls the mechanodynamics of macropinocytosis.
Proceedings of the National Academy of Sciences of the United States of America (2023-02-15)
Joseph Puccini, Jia Wei, Liang Tong, Dafna Bar-Sagi
ABSTRACT
Macropinocytosis is an actin-dependent mode of nonselective endocytosis that mediates the uptake of extracellular fluid-phase cargoes. It is now well recognized that tumor cells exploit macropinocytosis to internalize macromolecules that can be catabolized and used to support cell growth and proliferation under nutrient-limiting conditions. Therefore, the identification of molecular mechanisms that control macropinocytosis is fundamental to the understanding of the metabolic adaptive landscape of tumor cells. Here, we report that the acetyl-CoA-producing enzyme, ATP citrate lyase (ACLY), is a key regulator of macropinocytosis and describes a heretofore-unappreciated association of ACLY with the actin cytoskeleton. The cytoskeletal tethering of ACLY is required for the spatially defined acetylation of heterodimeric actin capping protein, which we identify as an essential mediator of the actin remodeling events that drive membrane ruffling and macropinocytosis. Furthermore, we identify a requirement for mitochondrial-derived citrate, an ACLY substrate, for macropinocytosis, and show that mitochondria traffic to cell periphery regions juxtaposed to plasma membrane ruffles. Collectively, these findings establish a mode of metabolite compartmentalization that supports the spatiotemporal modulation of membrane-cytoskeletal interactions required for macropinocytosis by coupling regional acetyl-CoA availability with dynamic protein acetylation.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid