Merck
  • Human Wharton's jelly-derived mesenchymal stem cells prevent pregnancy loss in a rat by JAK/STAT-mediated immunomodulation.
Human Wharton's jelly-derived mesenchymal stem cells prevent pregnancy loss in a rat by JAK/STAT-mediated immunomodulation.
The journal of obstetrics and gynaecology research (2023-07-19)
Xiaoying Ding, Rongrong Wu, Beibei Jin, Chunyu Zhu, Yuquan Zhang, Xiaoqing Yang
ABSTRACT
Spontaneous abortion (SA) is a multiple-original syndrome with immune imbalance as one of its major risk factors. As Wharton's jelly-mesenchymal stem cells (WJ-MSCs) are considered to be able to prevent abortion, this study aims to explore the currently poorly understood underlining molecular signaling pathways and regulatory mechanisms of WJ-MSCs in pregnancy maintenance. Abortion mode is established by subcutaneous injection of bromocriptine in rat on day 9 and abortion prevention is achieved by WJ-MSCs injection via tail vein. WJ-MSCs were cultured with/without the inhibitors of JAK/STAT or NF-κB. The uterus was collected on the 14th day of gestation and the rate of embryo absorption was calculated. The expression of Th1/Th2/Th3 cytokines in decidual, placental tissue, and peripheral blood was analyzed. WJ-MSCs treatment significantly reduced the abortion rate in bromocriptine-treated pregnancy such that it was not significantly different from a normal pregnancy. JAK/STAT inhibition abolished pregnancy preserving effects of WJ-MSCs but NF-κB inhibition did not. The levels of Th1-related cytokines and mRNA levels in the bromocriptine abortion model were significantly higher than the normal pregnancy group and ethanol control group, while levels of the Th2-related cytokines and mRNA levels significantly decreased. WJ-MSCs transfusion into the abortion model restored cytokine profiles such that they were not significantly different from the normal pregnancy group and ethanol control group. JAK/STAT inhibition of WJ-MSCs prevented their effect on cytokine and mRNA levels, but NF-κB inhibition did not. WJ-MSCs significantly lower the rate of embryo resorption of spontaneous abortion by reducing Th1-related cytokines while increasing Th2 and Th3-related cytokines in JAK/STAT-dependent manner.