Merck
  • Mural Wnt/β-catenin signaling regulates Lama2 expression to promote neurovascular unit maturation.
Mural Wnt/β-catenin signaling regulates Lama2 expression to promote neurovascular unit maturation.
Development (Cambridge, England) (2022-09-14)
Saptarshi Biswas, Sanjid Shahriar, Nicholas P Giangreco, Panos Arvanitis, Markus Winkler, Nicholas P Tatonetti, William J Brunken, Tyler Cutforth, Dritan Agalliu
ABSTRACT
Neurovascular unit and barrier maturation rely on vascular basement membrane (vBM) composition. Laminins, a major vBM component, are crucial for these processes, yet the signaling pathway(s) that regulate their expression remain unknown. Here, we show that mural cells have active Wnt/β-catenin signaling during central nervous system development in mice. Bulk RNA sequencing and validation using postnatal day 10 and 14 wild-type versus adenomatosis polyposis coli downregulated 1 (Apcdd1-/-) mouse retinas revealed that Lama2 mRNA and protein levels are increased in mutant vasculature with higher Wnt/β-catenin signaling. Mural cells are the main source of Lama2, and Wnt/β-catenin activation induces Lama2 expression in mural cells in vitro. Markers of mature astrocytes, including aquaporin 4 (a water channel in astrocyte endfeet) and integrin-α6 (a laminin receptor), are upregulated in Apcdd1-/- retinas with higher Lama2 vBM deposition. Thus, the Wnt/β-catenin pathway regulates Lama2 expression in mural cells to promote neurovascular unit and barrier maturation.
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone MA6, clone MA6, Chemicon®, from rat
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat