Merck
  • Replication dynamics identifies the folding principles of the inactive X chromosome.
Replication dynamics identifies the folding principles of the inactive X chromosome.
Nature structural & molecular biology (2023-08-11)
Rawin Poonperm, Saya Ichihara, Hisashi Miura, Akie Tanigawa, Koji Nagao, Chikashi Obuse, Takashi Sado, Ichiro Hiratani
ABSTRACT
Chromosome-wide late replication is an enigmatic hallmark of the inactive X chromosome (Xi). How it is established and what it represents remains obscure. By single-cell DNA replication sequencing, here we show that the entire Xi is reorganized to replicate rapidly and uniformly in late S-phase during X-chromosome inactivation (XCI), reflecting its relatively uniform structure revealed by 4C-seq. Despite this uniformity, only a subset of the Xi became earlier replicating in SmcHD1-mutant cells. In the mutant, these domains protruded out of the Xi core, contacted each other and became transcriptionally reactivated. 4C-seq suggested that they constituted the outermost layer of the Xi even before XCI and were rich in escape genes. We propose that this default positioning forms the basis for their inherent heterochromatin instability in cells lacking the Xi-binding protein SmcHD1 or exhibiting XCI escape. These observations underscore the importance of 3D genome organization for heterochromatin stability and gene regulation.