Merck
  • N-linked glycosylation is essential for anti-tumor activities of KIAA1324 in gastric cancer.
N-linked glycosylation is essential for anti-tumor activities of KIAA1324 in gastric cancer.
Cell death & disease (2023-08-24)
Rebecca Yun, Eunji Hong, Junil Kim, Bora Park, Staci Jakyong Kim, Bona Lee, Yong Sang Song, Seong-Jin Kim, Sujin Park, Jin Muk Kang
ABSTRACT
KIAA1324 is a transmembrane protein largely reported as a tumor suppressor and favorable prognosis marker in various cancers, including gastric cancer. In this study, we report the role of N-linked glycosylation in KIAA1324 as a functional post-translational modification (PTM). Loss of N-linked glycosylation eliminated the potential of KIAA1324 to suppress cancer cell proliferation and migration. Furthermore, we demonstrated that KIAA1324 undergoes fucosylation, a modification of the N-glycan mediated by fucosyltransferase, and inhibition of fucosylation also significantly suppressed KIAA1324-induced cell growth inhibition and apoptosis of gastric cancer cells. In addition, KIAA1324-mediated apoptosis and tumor regression were inhibited by the loss of N-linked glycosylation. RNA sequencing (RNAseq) analysis revealed that genes most relevant to the apoptosis and cell cycle arrest pathways were modulated by KIAA1324 with the N-linked glycosylation, and Gene Regulatory Network (GRN) analysis suggested novel targets of KIAA1324 for anti-tumor effects in the transcription level. The N-linked glycosylation blockade decreased protein stability through rapid proteasomal degradation. The non-glycosylated mutant also showed altered localization and lost apoptotic activity that inhibits the interaction between GRP78 and caspase 7. These data demonstrate that N-linked glycosylation of KIAA1324 is essential for the suppressive role of KIAA1324 protein in gastric cancer progression and indicates that KIAA1324 may have anti-tumor effects by targeting cancer-related genes with N-linked glycosylation. In conclusion, our study suggests the PTM of KIAA1324 including N-linked glycosylation and fucosylation is a necessary factor to consider for cancer prognosis and therapy improvement.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
SP600125, ≥98% (HPLC)