Merck
  • Multiplexed Biosensing of Proteins and Virions with Disposable Plasmonic Assays.
Multiplexed Biosensing of Proteins and Virions with Disposable Plasmonic Assays.
ACS sensors (2023-08-23)
Stephanie Wallace, Martin Kartau, Tarun Kakkar, Chris Davis, Agnieszka Szemiel, Iliyana Samardzhieva, Swetha Vijayakrishnan, Sarah Cole, Giuditta De Lorenzo, Emmanuel Maillart, Kevin Gautier, Adrian J Lapthorn, Arvind H Patel, Nikolaj Gadegaard, Malcolm Kadodwala, Edward Hutchinson, Affar S Karimullah
ABSTRACT
Our growing ability to tailor healthcare to the needs of individuals has the potential to transform clinical treatment. However, the measurement of multiple biomarkers to inform clinical decisions requires rapid, effective, and affordable diagnostics. Chronic diseases and rapidly evolving pathogens in a larger population have also escalated the need for improved diagnostic capabilities. Current chemical diagnostics are often performed in centralized facilities and are still dependent on multiple steps, molecular labeling, and detailed analysis, causing the result turnaround time to be over hours and days. Rapid diagnostic kits based on lateral flow devices can return results quickly but are only capable of detecting a handful of pathogens or markers. Herein, we present the use of disposable plasmonics with chiroptical nanostructures as a platform for low-cost, label-free optical biosensing with multiplexing and without the need for flow systems often required in current optical biosensors. We showcase the detection of SARS-CoV-2 in complex media as well as an assay for the Norovirus and Zika virus as an early developmental milestone toward high-throughput, single-step diagnostic kits for differential diagnosis of multiple respiratory viruses and any other emerging diagnostic needs. Diagnostics based on this platform, which we term "disposable plasmonics assays," would be suitable for low-cost screening of multiple pathogens or biomarkers in a near-point-of-care setting.
Sigma-Aldrich
Anti-CDK1/CDC2 Antibody (C-Term), serum, Upstate®